• USA Home
  • Z361356 - Slide recording system, digital palette
EMAIL THIS PAGE TO A FRIEND
Z361356 Sigma
Slide recording system, digital palette
RasterPlus P/S
Purchase
Properties
Related Categories Labware, Light Microscopy, Manufacturer Browser, Microscopy & Staining, Polaroid,
mfr. no.   Polaroid
availability   not available in EU
Description
Features and Benefits
Make top-quality 35mm color slides from any computer graphic. Printing to color transparency film is as easy as producing a black laser-printed page; these units are compatible with virtually all presentation graphics software and scanned-image viewers. Expose simple word slides in as little as 90 seconds, full-resolution complex graphics in about 4 minutes (depending on computer speed). The camera back accepts standard E-6 color slide film as well as Polaroid′s unique instant slide films that can be developed on-site in four minutes or less. Each unit comes with 3 rolls of Presentation Chrome E-6 color slide film. See the Photography section for films, processing chemicals, and Polaroid slide processors.
Windows-compatible PostScript interpreter; includes file translators for SCODL, PCX, and GIF file formats and 100 professionally designed backgrounds on which you can superimpose your images.
Price and Availability
Tissue Diagnostics
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?