• USA Home
  • Z361836 - Semi-dry blotters, W.E.P.
EMAIL THIS PAGE TO A FRIEND
Z361836 Sigma
Semi-dry blotters, W.E.P.
With photoelectric safety switch, AC input 120 V / 240 V
Purchase
Properties
Related Categories Blotters, Electrophoresis Supplies, Labware, Semi-Dry Blotters
Description
General description
Novel design incorporates several safety and convenience features. Proprietary resin-impregnated carbon electrodes deliver extremely uniform current and efficiently dissipate heat. The cover can only be placed in the proper orientation and the anode lead can be attached only with the cover in place.
Except for the last unit below, units have 18 × 18 cm electrode surface and accommodate transfer stacks up to six layers of gels and membranes (multi-gel stacks are not recommended for nucleic acid transfers). Units with safety switch and/or timer operate on 120/240 V, but 240 V line cords are not included.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?