• USA Home
  • Z361976 - Prism® for Windows®
EMAIL THIS PAGE TO A FRIEND
Z361976 Sigma
Prism® for Windows®
combination of biostatistics, curve fitting and scientific graphing
Purchase
Properties
Related Categories Computer Software, Data Analysis software, Labware More...
Description
General description
Prism is a powerful combination of biostatistics, curve fitting (nonlinear regression) and scientific graphing in one comprehensive program. Easily organize, analyze and graph repeated experiments; pick appropriate statistical tests and interpret the results. Seamless data analysis and full-featured graphing in one integrated program. You control the structure of your data table: store replicate determinations, related data sets, and repeated experiments all together; analyze all the data at once.
Enter data directly or import ASCII files or data from other Windows programs. Menu-driven data analysis: select one of the built-in nonlinear regression models or define your own. Versatile graphing allows you to define all aspects of your graph design and save your choices with the data set. Context-sensitive help offers suggestions and background theory on data analyses, explains calculations, and lets you customize elements of a graph by clicking on them.
System Requirements
Windows® 2000, XP or Vista.
Legal Information
Prism is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
Windows is a registered trademark of Microsoft Corporation
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?