• USA Home
  • Z362123 - Fit−Regression Analysis
EMAIL THIS PAGE TO A FRIEND
Z362123 Sigma
Fit−Regression Analysis
MacIntosh version
Purchase
Properties
availability   not available in EU
Description
General description
This comprehensive Excel macro performs linear and non-linear regressions with as many as six parameters. Results can be plotted in several different formats, including scatter plots with error bars.
System Requirements
Windows 3.1, 95, 98 or NT and Excel 4.0 or higher.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?