• USA Home
  • Z362743 - Nalgene® polycarbonate Erlenmeyer flask
EMAIL THIS PAGE TO A FRIEND
Z362743 Sigma
Nalgene® polycarbonate Erlenmeyer flask
capacity 500 mL
Synonym: conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer
Purchase
Properties
Related Categories Erlenmeyer Flasks, Plastic, Flasks, Labware, Manufacturer Browser, Nalgene,
material   polycarbonate
  wide-neck
sterility   non-sterile
feature   closure type Stoppers
packaging   case of 12
  pack of 4
mfr. no.   Nalgene, 4103
capacity   500 mL
Description
General description
Autoclavable; however, repeated autoclaving may cause some loss of mechanical strength.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?