• USA Home
  • Z362883 - Nalgene® benchtop β waste container
EMAIL THIS PAGE TO A FRIEND
Z362883 Sigma
Nalgene® benchtop β waste container
capacity 1,000 mL
Purchase
Description
Compatibility
Without inner bottle, container also accommodates standard 8 × 12 in. solid waste disposal bags.
General description
3/8 in. thick, encases waste bottle without obstructing access. Shield cap fits securely with bottle closure in place. Provided with wide-mouth polyethylene bottle suitable for aqueous solution. Disposal of organic solvents may require a different inner bottle.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?