• USA Home
  • Z363049 - Carestream® Kodak® BioMax® MS film
EMAIL THIS PAGE TO A FRIEND
Z363049 Sigma
Carestream® Kodak® BioMax® MS film
size 9.5 in. (24 cm) × 12 in. (30 cm)
Synonym: autoradiography film, xray film
Purchase
Properties
Related Categories Autoradiography, Autoradiography Film, Carestream Kodak, Carestream Kodak Autoradiography, Carestream Kodak BioMax MS film,
mfr. no.   Kodak MS-1 832 6886
size   9.5 in. (24 cm) × 12 in. (30 cm)
Description
Frequently Asked Questions
Frequently Asked Questions are available for this Product.
General description
Double-emulsion film specifically engineered for biotechnology applications. Provides maximum sensitivity for detection of 32P and 125I in Northern and Southern blots, reducing quantity of isotope needed.
Sheets of film with no interleaved paper
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?