• USA Home
  • Z363081 - Carestream® Kodak® BioMax® MS film
EMAIL THIS PAGE TO A FRIEND
Z363081 Sigma
Carestream® Kodak® BioMax® MS film
size 14 in. (35 cm) × 17 in. (43 cm)
Synonym: autoradiography film, xray film
Purchase
Properties
Related Categories Autoradiography, Autoradiography Film, Carestream Kodak, Carestream Kodak Autoradiography, Carestream Kodak BioMax MS film,
mfr. no.   Kodak MS-2 176 4505
size   14 in. (35 cm) × 17 in. (43 cm)
Description
Frequently Asked Questions
Frequently Asked Questions are available for this Product.
General description
Double-emulsion film specifically engineered for biotechnology applications. Provides maximum sensitivity for detection of 32P and 125I in Northern and Southern blots, reducing quantity of isotope needed.
Each sheet of film is sealed in a light-tight envelope
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Frequently Asked Questions
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis. These documents are located on the product detail page under Useful Links & Tools. Click on the following link to search for a Certificate of Analysis. Please click the following link to see the details on our Product Dating Information.
How do I get lot-specific information or a Certificate of Analysis?
A Certificate of Analysis is available by lot number and can be obtained through our Advanced Search Option: http://www.sigmaaldrich.com/catalog/AdvancedSearchPage.do
How do I find price and availability?
There are several ways to find pricing and availability for our products.  Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers. 
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS. To access the shipping information for this material, use the link on the product detail page for the product, or search here. 
My question is not addressed here, how can I contact Technical Service for assistance?
Use the option to the right to "Ask a Question" by email of a Technical Service Scientist.
How do I handle autoradiography film?
Please view autoradiography film tips provided by Carestream Health, Inc. http://www.carestream.com/autoradiography-film-tips.html
Show more questions
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?