• USA Home
  • Z363200 - Carestream® Kodak® BioMax® MS Intensifying Screens without lead blockers
EMAIL THIS PAGE TO A FRIEND
Z363200 Sigma
Carestream® Kodak® BioMax® MS Intensifying Screens without lead blockers
For 12 × 16 in.(30 x 40 cm) film
Synonym: autoradiography cassettes, xray film cassettes
Purchase
Description
General description
Phosphor emits green light, closely matched to the spectral sensitivity of BioMax MS film; provides highest sensitivity for 32P and 125I autoradiography.
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?