• USA Home
  • Z363502 - Scienceware® exposure cassette security bag
EMAIL THIS PAGE TO A FRIEND
Z363502 Sigma
Scienceware® exposure cassette security bag
pkg of 1 ea
Purchase
Description
Features and Benefits
Augmented light protection of exposure cassettes which may leak light when old, worn, or damaged. Made of black nylon with hook-and-loop sealing strip on flap. Accepts exposure cassettes up to 14 × 17 in. (35 × 43 cm).
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?