• USA Home
  • Z363642 - Barnstead/Thermolyne® Dewars, Biocane series
EMAIL THIS PAGE TO A FRIEND
Z363642 Sigma
Barnstead/Thermolyne® Dewars, Biocane series
Cart for transporting Dewars safely
Purchase
Properties
mfr. no.   Barnstead International AY509X9
Description
Compatibility
Accepts Biocane 20, 34, and 47
General description
A can and cane cryopreservation system with features to meet the requirements of almost any research lab: available in five sizes for small- to large-scale sample storage; unsurpassed temperature uniformity with low liquid nitrogen consumption rates; secure locking hasp prevents unauthorized access; each Biocane vessel includes six stainless steel canisters that accommodate Nalgene CryoCanes or similar storage systems. CryoCanes not included.
Legal Information
Bio-Cane is a trademark of Thermo Fisher Scientific or its subsidiaries
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?