• USA Home
  • Z363839 - Transilluminator UV/white light
EMAIL THIS PAGE TO A FRIEND
Z363839 Sigma
Transilluminator UV/white light
AC input 220 V AC50 Hz
Purchase
Description
General description
One transilluminator with two functions. The left side is a 20 × 20 cm white light table for reading autoradiograms, protein gels, and multiwell plates. It is uniformly illuminated by two 8W color-balanced fluorescent tubes diffused through an opal glass stage. The right side is a 20 × 20 cm, 312 nm UV transilluminator for ethidium bromide-stained gels and other fluorescence applications. Illumination comes from six 8W tubes. The UV filter resists solarization indefinitely with 312 nm light source. A hinged UV safety screen protects the entire stage and can be propped up like a work shield.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?