• USA Home
  • Z363944 - Chamber/Enclosure, UV
EMAIL THIS PAGE TO A FRIEND
Z363944 Sigma
Chamber/Enclosure, UV
AC input 220 V AC50 Hz
Purchase
Properties
Related Categories Labware, Molecular Biology Supplies, PCR (Polymerase Chain Reaction) Supplies, PCR Workstation
mfr. no.   Plas-Labs 825-UVC/220
Description
General description
Price includes shipping
Multi-purpose benchtop work enclosure for any procedure that can be affected by airborne contamination − culture work, DNA sequencing, PCR, etc. Removable side access panels provide a "still air" enclosure with both fluorescent and germicidal UV lamps. UV is automatically switched off whenever an access panel is removed. Front panel is 1/2-inch, sides and back are 3/8-inch acrylic for all-around β-protection and optical clarity. Two 6-inch wide white acrylic shelves can be installed in multi-position brackets at back of chamber. Electrical housing is removable for installation of large pieces of equipment. Dimensions of 24 in × 18 in (w × d) provide roomy working area. Unit is 28 in high.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?