• USA Home
  • Z364231 - Glycobiology: A Practical Approach
EMAIL THIS PAGE TO A FRIEND
Z364231 Sigma
Glycobiology: A Practical Approach
  •  ISBN-10 0-19-963371-1
  •  ISBN-13 978-0-19-963371-5
Purchase
Properties
publication info   D. Rickwood and B.D. Hames, ed., IRL Press at Oxford University Press, Oxford, England, 1993, 390 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
Describes the essential analytical techniques to study the structure and biosynthesis of glycoproteins.
Table of Contents
1. Isolation and characterization of glycoproteins
2. Compositional analysis of glycoproteins
3. Isolation and fractionation of N-linked oligosaccharides from glycoproteins
3a. Release of N-linked oligosaccharides and fractionation by gel filtration
3b. Fractionation of oligosaccharides by serial affinity chromatography with use of immobilized lectin columns
3c. Fractionation of glycopeptides and oligosaccharides from glycoproteins by HPLC
4. Structural characterization of oligosaccharides from glycoproteins
4a. Glycosidase treatment and other methods, including methylation analysis
4b. FAM-MS and ES-MS of glycoproteins
4c. Nuclear magnetic resonance spectroscopy of glycoprotein-derived carbohydrate chains
5. Structural characterization of N-glycans obtained from metabolically-radiolabelled glycoproteins
6. Structural characterization of mucin-type O-linked oligosaccharides
7. Characterization of nuclear and cytoplasmic glycoproteins
8. GPI membrane anchors
Appendix
Index
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?