• USA Home
  • Z365157 - Membrane filters for epifluorescent microscopy
EMAIL THIS PAGE TO A FRIEND
Z365157 Sigma
Membrane filters for epifluorescent microscopy
pore size 0.4 μm, diam. 25 mm
Purchase
Properties
Related Categories Electron Microscopy, Electron Microscopy Epifluorescent Filters, Labware, Microscopy & Staining
material   black polycarbonate
diam.   25 mm
pore size   0.4 μm
Description
General description
Track-etch membranes provide a flat, smooth surface with highly uniform pores, an ideal medium for collecting particles for microscopic analysis. These membranes are especially designed for epifluorescent microscopy of microorganisms in water and microbiological media. The blackening process eliminates natural autofluorescence and provides a high-contrast background for observing fluorescent-stained microbes.
Price and Availability
Tissue Diagnostics
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?