• USA Home
  • Z365408 - Eppendorf® Reference® series adjustable-volume pipettes
EMAIL THIS PAGE TO A FRIEND
Z365408 Sigma
Eppendorf® Reference® series adjustable-volume pipettes
volume 0.5-10 μL
Purchase
Properties
Related Categories Eppendorf, Eppendorf Pipettors, Eppendorf Pipettors & Tips, Labware, Liquid Handling Equipment,
feature   autoclavable
  electronic: no
  single-channel
mfr. no.   Eppendorf®, 022470051 (USA)
  Eppendorf®, 4910 000.018 (European)
  Eppendorf®, 4910 000.018
volume   0.5-10 μL
Description
General description
Locking volume-adjustment button: push in for easy volume change; release to lock setting. Shorter length for easier handling, especially in hoods. Single-button control of filling, delivery, and tip ejection. Ejector sleeve is removable for access to narrow vessels.
Other Notes
Ultra-Micro
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?