EMAIL THIS PAGE TO A FRIEND
Z365793 Sigma
TableCurve
2D for Windows
Purchase
Properties
Related Categories Computer Software, Data Analysis software, Labware More...
mfr. no.   AISN
  Spss
Description
Compatibility
• System Requirements: Windows 95, 98, 2000, NT
• Pentium™ or compatible processor
• 32 MB random-access memory (RAM). NOTE: 64 MB RAM is recommended for processing large data sets and for the automation procedures
• 20 MB of available disk space
• 800 × 600 resolution (SVGA) or higher and 256 colors
General description
Automate the difficult task of finding the best equation to fit experimental data. Import data as ASCII files or from Lotus, Excel, or Quattro Pro spreadsheets. Then TableCurve, in a single processing step, fits the data to all of its built-in equations and ranks them by correlation coefficient. View results as graphs or numeric summaries. Output hardcopy graphs or create a file of fit results. Choose TableCurve 2D if your data involves curve-fitting two-variable (x,y) data; TableCurve 3D for surface-fitting three-variable (x,y,z) data.
Legal Information
TableCurve is a trademark of AISN Software
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?