• USA Home
  • Z366048 - Eppendorf® Microcentrifuge accessories
EMAIL THIS PAGE TO A FRIEND
Z366048 Sigma
Eppendorf® Microcentrifuge accessories
Fixed Angle Rotor with lid, (Rotor F-45-30-11), for use with 5417 Microcentrifuge
Purchase
Properties
Related Categories Centrifugation Supplies, Centrifuges, Eppendorf, Eppendorf Centrifugation, Eppendorf Centrifuge parts & Accessories,
material   aluminum lid
mfr. no.   Eppendorf®, 022636006 (USA)
  Eppendorf®, 5490 015.002 (European)
Description
General description
30 × 11 mm rotor bores for 1.5/2.0 mL microcentrifuge tubes
Max 14,000 rpm, max. rcf 20,800 g
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Steriflip Filters Units
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?