• USA Home
  • Z366102 - Eppendorf® Microcentrifuge accessories
EMAIL THIS PAGE TO A FRIEND
Z366102 Sigma
Eppendorf® Microcentrifuge accessories
Aerosol-tight capsule kit
Purchase
Properties
mfr. no.   Eppendorf®
  Eppendorf®
Description
Compatibility
For Model 5417
Provides individual containment of microcentrifuge tubes. Fit only in 24-place fixed angle rotor above. Kit includes 12 capsule bodies, 20 springs, 100 caps, and 2 racks.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?