• USA Home
  • Z367672 - Hybridization roller bottles
EMAIL THIS PAGE TO A FRIEND
Z367672 Sigma
Hybridization roller bottles
size 29 cm × 21.9 cm
Purchase
Properties
Related Categories Hybridization Supplies, Hybridization Tubes, Labware, Molecular Biology Supplies
size   29 cm × 21.9 cm
Description
Features and Benefits
Will accommodate membranes from strips to large blots. Bottles are 1/4′′ thick acrylic. Dimensions given are inside length × inside circumference; as such, they correspond to the size of the flat membrane that can fit in without overlap.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?