• USA Home
  • Z367869 - Scienceware® colony counter system
EMAIL THIS PAGE TO A FRIEND
Z367869 Sigma
Scienceware® colony counter system
Mini light box
Purchase
Properties
Related Categories Cell Culture Supplies, Colony Counting, Labware More...
feature   CE compliant
mfr. no.   Bel-Art
battery power   4 AA (not included)
Description
General description
Powered by 4 AA batteries (not included); the power AC adapter BAH378640301 is NOT included. A removable counting grid is included. Dimensions: 158.8 X 136.5 X 44.5mm H (61/4 X 53/8 X 13/4"). Weighs 255 g (9oz).
Color-rectified fluorescent observation surface
• Perfect for slide, plate, mini-gel or x-ray film analysis
• Compact, lightweight system
• Includes a counting grid that can be removed
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Scepter 2.0
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?