• USA Home
  • Z367990 - Cellulose acetate membrane

EMAIL THIS PAGE TO A FRIEND
Z367990 Sigma

Cellulose acetate membrane

dialyser capacity 10-200 μL, MWCO 1,000 Da, diam. 11 mm

Purchase

Properties

Related Categories Biodialyser, Dialysis, Labware More...
packaging   pkg of 25 membranes
  pkg of 25 pkg
dialyser capacity   10-200 μL
diam.   11 mm
pore size   1,000 Da MWCO
storage temp.   2-8°C

Description

General description

Made of cellulose acetate for low protein binding. Nominal molecular weight limit (MWCO) is approximate upper limit of molecular weight to which membrane is permeable.

Price and Availability


Mitex PTFE Membrane Filters
Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?