Z368032 Sigma

Cellulose acetate membrane

diam. 16 mm, MWCO 1,000 Da

DOWNLOAD MSDS (PDF)

Properties

Related Categories Biodialyser, Dialysis, Labware More...
packaging   pkg of 25 membranes
diam.   16 mm
pore size   1,000 Da MWCO

Description

Compatibility

for 500-1500 μL chamber

General description

Made of cellulose acetate for low protein binding. Nominal molecular weight limit (MWCO) is approximate upper limit of molecular weight to which membrane is permeable.

Price and Availability

Customers Also Viewed

Biodialysers

Double sided, sample reservoir size 1,000 μL

Cellulose acetate membrane

dialyser capacity 10-200 μL, MWCO 500 Da, diam. 11 mm

Cellulose acetate membrane

dialyser capacity 500-1,500 μL, MWCO 500 Da, diam. 16 mm

Cellulose acetate membrane

dialyser capacity 10-200 μL, MWCO 1,000 Da, diam. 11 mm

Dispo-Biodialyzer

MWCO 1 kDa

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?