• USA Home
  • Z368059 - Cellulose acetate membrane

EMAIL THIS PAGE TO A FRIEND
Z368059 Sigma

Cellulose acetate membrane

dialyser capacity 500-1,500 μL, MWCO 50,000 Da, diam. 16 mm

Purchase

Properties

Related Categories Biodialyser, Dialysis, Labware More...
packaging   pkg of 25 membranes
  pkg of 25 pkg
dialyser capacity   500-1,500 μL
diam.   16 mm
pore size   50,000 Da MWCO
storage temp.   2-8°C

Description

General description

Made of cellulose acetate for low protein binding. Nominal molecular weight limit (MWCO) is approximate upper limit of molecular weight to which membrane is permeable.

Price and Availability


Mitex PTFE Membrane Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?