• USA Home
  • Z368075 - Eppendorf® Research® pipette, adjustable volume
EMAIL THIS PAGE TO A FRIEND
Z368075 Sigma
Eppendorf® Research® pipette, adjustable volume
volume 0.1-2.5 μL
Purchase
Properties
Related Categories Eppendorf, Eppendorf Pipettors, Eppendorf Pipettors & Tips, Labware, Liquid Handling Equipment,
feature   electronic: no
  single-channel
mfr. no.   Eppendorf®, 022471856 (USA)
  Eppendorf®, 3111 000.114 (European)
volume   0.1-2.5 μL
Description
General description
If you prefer a separate tip ejector button, choose the Series 2100. Easy-turn volume adjustment moves quickly through the volume range and won′t snag gloves. Integrated ejector sleeve allows access to narrow vessels. Lower section is autoclavable. Ultra-micro models have shielded piston to protect against damage.
Other Notes
Ultra-Micro
Legal Information
Eppendorf Research is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z683779 Eppendorf® Research® plus pipette, variable volume, volume 0.1-2.5 μL
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?