• USA Home
  • Z368180 - Eppendorf® centrifugal vacuum concentrator
EMAIL THIS PAGE TO A FRIEND
Z368180 Sigma
Eppendorf® centrifugal vacuum concentrator
Basic Model 5301, AC input 220 V AC50 - 60 Hz
Purchase
Properties
Related Categories Centrifugation Supplies, Centrifuges, Eppendorf Centrifuges, Labware
mfr. no.   Eppendorf®, 5301 000.016 (European)
Description
General description
Basic unit has AC outlet for connection to your vacuum pump. Vacuum release is still controlled by the concentrator.
Concentrates aqueous and alcoholic solutions quickly and cleanly right in their microtube containers. The unit applies gentle heat and vacuum to evaporate the solvent, while centrifugation prevents bumping and foaming. Choice of four temperature settings (ambient, 30, 45, and 60 °C) allows processing of a variety of samples. Vacuum is automatically released before the centrifuge turns off. The 48-place rotor for 1.5-2 mL tubes and the 72-place rotor for 0.5 mL tubes can be stacked two high for double the throughput.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Steriflip Filters Units
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?