• USA Home
  • Z368210 - Eppendorf® centrifugal vacuum concentrator
EMAIL THIS PAGE TO A FRIEND
Z368210 Sigma
Eppendorf® centrifugal vacuum concentrator
Rotor for Model 5301, 48 place for 1.5-2 ml tubes
Purchase
Properties
quality   48 place for 1.5-2 ml tubes
mfr. no.   Eppendorf®
  Eppendorf®
Description
General description
Concentrates aqueous and alcoholic solutions quickly and cleanly right in their microtube containers. The unit applies gentle heat and vacuum to evaporate the solvent, while centrifugation prevents bumping and foaming. Choice of four temperature settings (ambient, 30, 45, and 60 °C) allows processing of a variety of samples. Vacuum is automatically released before the centrifuge turns off. The 48-place rotor for 1.5-2 mL tubes and the 72-place rotor for 0.5 mL tubes can be stacked two high for double the throughput.
Stackable two high; one rotor spacer (below) required
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?