• USA Home
  • Z368679 - Biodialyser fittings
EMAIL THIS PAGE TO A FRIEND
Z368679 Sigma
Biodialyser fittings
Solid end cap for converting 50-200 μl double-sided Biodialyser to single sided
Purchase
Properties
Related Categories Biodialyser, Dialysis, Labware More...
Description
Compatibility
Must also be used to replace one membrane and end cap in a double-sided Biodialyser for vacuum concentration after dialysis
Price and Availability
Mitex PTFE Membrane Filters
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?