• USA Home
  • Z368784 - Finnpipette® digital single channel pipette
EMAIL THIS PAGE TO A FRIEND
Z368784 Sigma
Finnpipette® digital single channel pipette
20-200 μL, resolution: 1.0 μL
Purchase
Properties
mfr. no.   Finn
measuring range   20-200 μL, resolution: 1.0 μL
Description
General description
New ergonomic design with all the best features of earlier Finnpipette lines: fully autoclavable without disassembly; certified factory calibration and easy in-lab recalibration (tool and instructions included); soft-touch tip ejection provides more ejector force with less thumb pressure.
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z678139 Finnpipette® F3 variable volume pipette, capacity 20-200 μL
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?