• USA Home
  • Z368881 - Finnpipette® digital fixed-volume pipettes
EMAIL THIS PAGE TO A FRIEND
Z368881 Sigma
Finnpipette® digital fixed-volume pipettes
size 200 μL
Purchase
Description
General description
Dedicated pipettes for repetitive single-volume pipetting. Ergonomic design reduces fatigue; soft-touch tip ejector provides more force with less thumb pressure; easy in-lab calibration; fully autoclavable without disassembly.
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?