• USA Home
  • Z368954 - Finnpipette® digital multichannel pipette
EMAIL THIS PAGE TO A FRIEND
Z368954 Sigma
Finnpipette® digital multichannel pipette
8 channel, size 5-50 μL , in 0.5 μL increment
Purchase
Description
General description
Eight- and twelve-channel pipettes from the pioneer in multichannel pipettes. Tip spacing allows quick filling of 96-well plates by columns or rows. Ergonomic design and light weight reduce fatigue. Certified factory calibration and easy in-lab recalibration. Gear-driven soft-touch ejector for de-tipping with less thumb pressure. Fully autoclavable.
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?