Synonym: autoradiography film, xray film
| Related Categories | Autoradiography, Autoradiography Film, Carestream Kodak, Carestream Kodak Autoradiography, Carestream Kodak BioMax light film, |
| mfr. no. | Kodak Light-2 Ready Pack 191 7012 |
| size | 5 in. (13 cm) × 7 in. (18 cm) |
Double-emulsion film on clear base. Provides clearest background and highest sensitivity for chemiluminescence detection systems. Also suitable for autoradiography, but with less sensitivity for high-energy isotopes than X-Omat AR and BioMax MS, and than BioMax MR for medium-energy isotopes.
Each sheet individually wrapped in a light-tight envelope
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Customers Also Viewed
pack of 1 gal