• USA Home
  • Z370487 - Easi-Grid master plating device
EMAIL THIS PAGE TO A FRIEND
Z370487 Sigma
Easi-Grid master plating device
Spare set of Easi-Grid patterns
Synonym: plating dish, plating dishes
Purchase
Properties
Related Categories Cell Culture Supplies, Labware, Master Plating More...
mfr. no.   Jencons
Description
General description
Essential tool for master plate making. Holds any 90-100 mm petri dish firmly in place above any of nine popular grid patterns for molecular biology, genetics, and microbiology applications. Grids are printed on clear acrylic, so they can be used on benchtop or light box. Patterns include 25, 50, 70, 100, 150, and 200 squares; 6 and 12 circle segments; a black 100-square grid; and a blank for custom grid design.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?