• USA Home
  • Z370924 - IEC Swinging bucket rotor aerocarriers
EMAIL THIS PAGE TO A FRIEND
Z370924 Sigma
IEC Swinging bucket rotor aerocarriers
capacity, 4 7-10–mL blood tubes
Purchase
Properties
feature   RCF 2200 × g
  capacity 4 7-10–mL blood tubes
  limits 3700 rpm
mfr. no.   IEC
availability   not available in EU
Description
Features and Benefits
Polypropylene buckets with clear lids and O-ring provide full aerosol containment. Lightweight, they speed up and slow down quickly. Clear caps allow inspection for breakage before opening. Aerocarriers can be autoclaved or decontaminated with a 1:10 dilution of bleach.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?