• USA Home
  • Z371211 - Finnpipette® BioControl 8-channel pipette
EMAIL THIS PAGE TO A FRIEND
Z371211 Sigma
Finnpipette® BioControl 8-channel pipette
volume range 5-50 μL, with U.S. plug
Purchase
Properties
Related Categories Finn Pipettors, Labware, Liquid Handling Equipment, Pipettes, Pipettors and Tips, Pipettors More...
mfr. no.   Finn
volume range   5-50 μL
Description
General description
The all-in-one liquid handling solution for the busy biotech lab. The overall system consists of a universal handle with recharger stand, and ten fully interchangeable tip cone modules. The handle incorporates these advanced features:
• natural trigger action pipetting
• power-assisted tip ejector with right- or left-handed operation
• three pipetting speeds for different viscosities
• forward and reverse modes, regular and stepping
• simple user calibration
• audible signal and flashing message indicate low battery
• rechargable with your choice of U.S., Euro, or U.K. recharger stands
Tip cones modules are fully autoclavable and accept standard Finntips. Multichannel modules rotate to user′s preferred position. Recharger automatically starts overnight recharging when pipette is placed on stand; rapid recharge (45-60 minutes) can be manually activated. Each BioControl system comes with multi-channel module, and includes recharger stand and calibration software. Update the system now or later with additional tip modules as your lab′s requirements dictate.
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z647098 BRAND® Transferpette® multichannel, electronic pipette, AC input 115 V, US/Japan plug, 12 channel, capacity 5-100 μL
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?