• USA Home
  • Z371661 - Staining tube Leighton
EMAIL THIS PAGE TO A FRIEND
Z371661 Sigma
Staining tube Leighton
sterile
Purchase
Properties
material   polymethylpentene slide (culture treated)
  polystyrene tube (optically clear; untreated)
sterility   sterile
availability   not available in EU
Description
General description
Screw-cap tube with self-contained microscope slide ideal for staining or fixing individual cultures.The slide is 9× 55mm. Supplied with screw caps. Tube has a frosted area for labeling
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?