• USA Home
  • Z372013 - Ohaus® Scout® balance

EMAIL THIS PAGE TO A FRIEND
Z372013 Sigma

Ohaus® Scout® balance

SP401, AC/DC input 120 V

Purchase

Properties

Related Categories Balances and Accessories, Labware, Ohaus Scout Balances More...
feature   CE compliant
  CSA approved
  display 0.7 in. (LCD)
  operating temperature 10 ‑ 40 °C
mfr. no.   SP401
availability   not available in EU
measuring range   400 g weighing capacity, resolution: 0.1 g
platform size   5 in. (12.7 cm) × 5 3/4 in. (14.6 cm)
battery power   (battery not included; AC adapter included)

Description

General description

Scout portable precision balances feature: simple two-button operation, easy calibration; weigh-below hook; sealed front panel; and spill ring; built-in security bracket.

Legal Information

Ohaus is a registered trademark of Ohaus Corp.

Scout is a registered trademark of Ohaus Corp.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?