• USA Home
  • Z372188 - Corning® microcentrifuge tubes with screw cap
EMAIL THIS PAGE TO A FRIEND
Z372188 Sigma
Corning® microcentrifuge tubes with screw cap
volume 1.5 mL, attached cap style
Purchase
Properties
Related Categories 1.0 to 2.0 mL microcentrifuge tubes, Centrifugation Supplies, Labware, Microcentrifuge Tubes
material   cap screw top
  tube conical bottom
  natural polypropylene tube
description   not self standing
sterility   sterile
feature   O-ring
  RCF 20,000 × g
mfr. no.   Corning 430909
packaging   pack of 50
  case of 500
volume   1.5 mL
Description
General description
Corning polypropylene microcentrifuge tubes feature screw caps that provide a tight, secure seal.
• Maximum RCF is 20,000
• Black silicone O-rings
• No marking spot
Legal Information
Corning is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?