• USA Home
  • Z372293 - Gel slider sequencing gel casting unit
EMAIL THIS PAGE TO A FRIEND
Z372293 Sigma
Gel slider sequencing gel casting unit
For gels 22−42cm long
Purchase
Description
General description
Use the sliding plate technique instead of unreliable, leak-prone tape, clamps, or plastic film. Gels are cast in minutes in a horizontal position, virtually eliminating leaks and trapped air bubbles. Just: (1) adjust the Gel Slider to the desired width (from 20 to 38 cm); (2) position the solid plate with spacers clamped to it; (3) position the top of the notched plate slightly over-lapping the bottom of the solid plate; (4) simultaneously add gel solution and slide the top plate over the bottom plate, removing clamps as you go; (5) when plates are flush, insert comb, clamp gel sandwich, and allow gel to polymerize.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?