• USA Home
  • Z372374 - Horizontal submarine electrophoresis units
EMAIL THIS PAGE TO A FRIEND
Z372374 Sigma
Horizontal submarine electrophoresis units
gel size 7.5 cm × 10 cm, base unit, dual mode, mini-gel 7.5 cm wide, green
Purchase
Properties
Related Categories Electrophoresis Supplies, Horizontal Gel Electrophoresis Supplies, Labware, Units and Components for Horizontal Gel Electrophoresis
material   acrylic
gel size   7.5 cm × 10 cm
color   green
Description
General description
Submarine gels may be formed directly on the UV-transparent gel bed using small sliding baffles to contain the agarose as it cools, or outside the unit using a UV-transparent gel tray sealed with tape. Gel is 7.5 cm wide × 10 cm long. Different colors are offered for any color-coding your lab requires (e.g., RNA only).
Combs and tape are not included. Gel tray is not included with the Mini-Gel unit.
Base unit, safety cover, power leads, and all other necessary parts are included.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?