• USA Home
  • Z372463 - Combs for vertical mini-gel, 11.3 cm wide
EMAIL THIS PAGE TO A FRIEND
Z372463 Sigma
Combs for vertical mini-gel, 11.3 cm wide
size 0.5 mm × 14 wells
Purchase
Description
General description
The following components are necessary to construct one complete single gel casting assembly: one set of glass plates, one set of spacers, one gasket, one comb, and gel casting clamps (one set of three for mini-size gel; two sets of three for standard size gels). Be sure to select the same length glass plates, spacers and gaskets; the same thickness spacers, comb and gasket.
For dual gel casting, order two of each component, but substitute dual gel casting clamps for the single gel casting clamps. For the second dimension of 2D electrophoresis, spacers, gaskets and a "1-well" comb must match the thickness of the first dimension tube gel.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?