• USA Home
  • Z373052 - Silicone cord replacement repair kit
EMAIL THIS PAGE TO A FRIEND
Z373052 Sigma
Silicone cord replacement repair kit
For standard vertical electrophoresis units (16.5 cm gel and wider)
Purchase
Description
Features and Benefits
Replaces the silicone sponge seal on the upper gel reservoir when it has been damaged or has lost its effectiveness with use. Includes all materials and instructions for replacement.
General description
Includes 48 in. of 3/16 in. white silicone cord, 1 tube of silicone adhesive with applicator tip and instructions.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?