• USA Home
  • Z373389 - Ohaus® Explorer® balance
EMAIL THIS PAGE TO A FRIEND
Z373389 Sigma
Ohaus® Explorer® balance
100-210 g weighing capacity, resolution: 0.1-1.0 mg, AC input 120 V, internal calibration: no
Purchase
Properties
Related Categories Balances and Accessories, Labware, Ohaus Explorer Balances More...
feature   CE compliant
  CSA approved
  display 7-character alphanumeric LCD for both weight readings and function selection
  internal calibration: no
  operating temperature 10 ‑ 40 °C (10-30 °C without internal calibration)
mfr. no.   Ohaus
availability   not available in EU
measuring range   100-210 g weighing capacity, resolution: 0.1-1.0 mg
free height above pan   279 mm
platform diam.   3 1/2 in. (90 mm)
Description
General description
Special features: full range tare; weighs in nine modes + custom units; bidirectional RS232 interface and remote/auxiliary port; application programs include GLP/GMP, parts counting, and animal weighing; modular design allows simple upgrade to Voyager series feature set.
Legal Information
Explorer is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?