• USA Home
  • Z373478 - Gel casting tray for horizontal submarine units
EMAIL THIS PAGE TO A FRIEND
Z373478 Sigma
Gel casting tray for horizontal submarine units
size 14 cm × 10 cm
Purchase
Description
Features and Benefits
Construction of 1/4 in. thick UV-transparent acrylic prevents distortion during gel formation and permits use on a UV transilluminator. Grooves provide for uniform placement of combs near either end or toward the center of the gel. Wells in base form agarose "anchors," preventing gel from floating. Extra trays permit pouring one gel while running another.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?