• USA Home
  • Z373524 - Carestream® Kodak® BioMax® light film
EMAIL THIS PAGE TO A FRIEND
Z373524 Sigma
Carestream® Kodak® BioMax® light film
size 12 in. (30 cm) × 16 in. (40 cm)
Purchase
Properties
mfr. no.   Light-1
size   12 in. (30 cm) × 16 in. (40 cm)
Description
General description
Double-emulsion film on clear base. Provides clearest background and highest sensitivity for chemiluminescence detection systems. Also suitable for autoradiography, but with less sensitivity for high-energy isotopes than X-Omat AR and BioMax MS, and than BioMax MR for medium-energy isotopes.
Sheets of film with no interleaved paper
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?