• USA Home
  • Z373885 - Sigma Rarex Blue III Intensifying Screens without lead blockers
EMAIL THIS PAGE TO A FRIEND
Z373885 Sigma
Sigma Rarex Blue III Intensifying Screens without lead blockers
For 14 × 17 in and 35 × 43 cm film
Purchase
Properties
Related Categories Autoradiography, Autoradiography Intensifying Screens, Electrophoresis Supplies, Labware
availability   not available in EU
Description
General description
A blue-emitting, rare earth screen which is compatible with full or half speed films. The construction of these screens is specially designed to maximize useful life and minimize damage due to moisture and other common contaminants.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?