• USA Home
  • Z374318 - Intensifying screens BioMax® Transcreen®
EMAIL THIS PAGE TO A FRIEND
Z374318 Sigma
Intensifying screens BioMax® Transcreen®
HE for 8 in. × 10 in. film
Synonym: autoradiography cassettes, xray film cassettes
Purchase
Description
General description
A new concept in intensifying screens provides improved resolution and maximum sensitivity and can now enhance exposures with low-energy β-emitters.
Conventional intensifying screens capture particles after they have passed through the film and "reflect" them back as light. Low-energy particles can never reach a conventional intensifying screen. TranScreens, placed between the gel or blot and the film, intercept particles and "transmit" them to the film as visible light. Choose from:
• LE for low-energy β-emitters like 3H, 14C, 33P, 35S, 45Ca, and 59Fe
• HE for high-energy β-emitters like 32P and γ-emitters like 125I
Both are optimized for use with BioMax MS film, but can be used with other films.
Can easily be cut to fit 18 × 24 cm film
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Transcreen is a registered trademark of Kodak
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?