• USA Home
  • Z374482 - β acrylic lock boxes, Nalgene®
EMAIL THIS PAGE TO A FRIEND
Z374482 Sigma
β acrylic lock boxes, Nalgene®
size 286 mm × 324 mm × 127 mm
Purchase
Properties
mfr. no.   Nalgene
availability   not available in EU
size   286 mm × 324 mm × 127 mm
Description
Features and Benefits
Small bench-top unit. Useful for refrigerator or freezer storage
General description
β acrylic lock boxes have no metal parts to produce secondary x-rays. They are made from 1/2in. (12mm) thick acrylic with built-in push-button combination locks.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?