• USA Home
  • Z374520 - Brinkmann bottle-top dispenser
EMAIL THIS PAGE TO A FRIEND
Z374520 Sigma
Brinkmann bottle-top dispenser
2.0-10.0 mL
Purchase
Properties
Related Categories Brinkmann Bottle-Top Dispenser, Dispensers, Labware, Liquid Handling Equipment
mfr. no.   Brinkman
availability   not available in EU
measuring range   2.0-10.0 mL
Description
General description
Brinkman bottletop dispensers offer a unique combination of features:
• PTFE "wiping" piston displaces reagent from the cylinder without leaving any residue behind, eliminating piston swelling and freezing
• Grooved positive lock volume adjustment
• Unique telescoping filling tube
• Individual calibration certificate
• Completely autoclavable without disassembly
• Clear plastic sleeve surrounds and protects the glass cylinder
Accuracy ±0.5% (±0.6% for 0.1-2.5 mL size). Reproducibility of 0.1%.
Adapters and accessories allow connection to almost any size reagent bottle. Dispensers with maximum volume of 2.5 to 10 mL connect directly to bottles with 33 mm threaded neck and are supplied with 28 mm, 38 mm, and 45 mm adapters. Dispensers with maximum volumes of 25 to 100 mL connect directly to bottles with 45 mm threaded neck and are supplied with 33 mm and 38 mm adapters
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?