• USA Home
  • Z374628 - Eppendorf® Combitips® Plus
EMAIL THIS PAGE TO A FRIEND
Z374628 Sigma
Eppendorf® Combitips® Plus
volume range 2.5 mL
Purchase
Properties
Related Categories Combitips Plus, Eppendorf, Eppendorf Pipette Tips, Eppendorf Pipettors & Tips, Eppendorf Positive Displacement,
material   colorless tip
sterility   non-sterile
feature   barrier: No
packaging   pack of 100
mfr. no.   Eppendorf®, 0030 069.242 (European)
  Eppendorf®, 022266306 (USA)
volume range   2.5 mL
Description
General description
Snaps securely into Multipette pipette. Tips are not autoclavable.
Legal Information
Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?